missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenyl isothiocyanate, 96%

Catalog No. p-7024934
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2020403 1 g
AAB2020406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2020403 Supplier Thermo Scientific Chemicals Supplier No. B2020403

CAS: 141106-52-7 | C7H3F2NS | 171.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 141106-52-7
Molecular Formula C7H3F2NS
Molecular Weight (g/mol) 171.165
MDL Number MFCD00041044
InChI Key ABGGPKIFVAIRGU-UHFFFAOYSA-N
Synonym 2,4-difluorophenyl isothiocyanate, 2,4-difluoro isothiocyanato benzene, 2,4-difluoro-1-isothiocyanato-benzene, 2,4-difluorophenylisothiocyanate, isothiocyanic acid 2,4-difluorophenyl ester, benzene, 2,4-difluoro-1-isothiocyanato, benzene, 2,4-difluoro-1-isothiocyanato-9ci, 2,4-difluorobenzenisothiocyanate, pubchem4451, acmc-209clq
PubChem CID 518879
IUPAC Name 2,4-difluoro-1-isothiocyanatobenzene
SMILES C1=CC(=C(C=C1F)F)N=C=S

Specifications

Density 1.349
Boiling Point 98°C to 99°C (30 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.597
Quantity 1 g
UN Number UN2922
Beilstein 6919744
Sensitivity Moisture sensitive
Formula Weight 171.17
Percent Purity 96%
Chemical Name or Material 2,4-Difluorophenyl isothiocyanate
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 624-035-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only