Learn More
2,4-Difluorophenyl isocyanate, 98+%
CAS: 59025-55-7 | C7H3F2NO | 155.10 g/mol
$84.56 - $760.85
Chemical Identifiers
| CAS | 59025-55-7 |
|---|---|
| Molecular Formula | C7H3F2NO |
| Molecular Weight (g/mol) | 155.10 |
| MDL Number | MFCD00001997 |
| InChI Key | HNENEALJPWJWJY-UHFFFAOYSA-N |
| Synonym | 2,4-difluorophenyl isocyanate, 2,4-difluorophenylisocyanate, benzene, 2,4-difluoro-1-isocyanato, isocyanic acid 2,4-difluorophenyl ester, 2,4-difluoro-1-isocyanato-benzene, 2,4-difluoroisocyanatobenzene, timtec-bb sbb006537, pubchem4453, 2,4-diflurophenylisocyanate, acmc-1b06k |
| PubChem CID | 593258 |
| IUPAC Name | 2,4-difluoro-1-isocyanatobenzene |
| SMILES | FC1=CC=C(N=C=O)C(F)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1241506
|
Thermo Scientific Chemicals
A1241506 |
5 g |
Each for $84.56
|
|
|||||
|
AAA1241514
|
Thermo Scientific Chemicals
A1241514 |
25 g |
Each for $264.79
|
|
|||||
|
AAA1241522
|
Thermo Scientific Chemicals
A1241522 |
100 g |
Each for $760.85
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 59025-55-7 | |
| 155.10 | |
| HNENEALJPWJWJY-UHFFFAOYSA-N | |
| 593258 | |
| FC1=CC=C(N=C=O)C(F)=C1 |
| C7H3F2NO | |
| MFCD00001997 | |
| 2,4-difluorophenyl isocyanate, 2,4-difluorophenylisocyanate, benzene, 2,4-difluoro-1-isocyanato, isocyanic acid 2,4-difluorophenyl ester, 2,4-difluoro-1-isocyanato-benzene, 2,4-difluoroisocyanatobenzene, timtec-bb sbb006537, pubchem4453, 2,4-diflurophenylisocyanate, acmc-1b06k | |
| 2,4-difluoro-1-isocyanatobenzene |
Specifications
| 59025-55-7 | |
| 64°C to 66°C (10 mmHg) | |
| C7H3F2NO | |
| MFCD00001997 | |
| UN3080 | |
| Moisture sensitive | |
| HNENEALJPWJWJY-UHFFFAOYSA-N | |
| 2,4-difluoro-1-isocyanatobenzene | |
| 593258 | |
| ≥98% |
| 1.31 | |
| 50°C (122°F) | |
| 1.492 | |
| 5 g | |
| 2831577 | |
| 2,4-difluorophenyl isocyanate, 2,4-difluorophenylisocyanate, benzene, 2,4-difluoro-1-isocyanato, isocyanic acid 2,4-difluorophenyl ester, 2,4-difluoro-1-isocyanato-benzene, 2,4-difluoroisocyanatobenzene, timtec-bb sbb006537, pubchem4453, 2,4-diflurophenylisocyanate, acmc-1b06k | |
| FC1=CC=C(N=C=O)C(F)=C1 | |
| 155.10 | |
| 155.11 | |
| 2,4-Difluorophenyl isocyanate |
Safety and Handling
GHS H Statement
H300-H330-H334-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 3; Packaging Group: II
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only