missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenyl isocyanate, 98+%

CAS: 59025-55-7 | C7H3F2NO | 155.10 g/mol

Supplier:  Thermo Scientific Chemicals A1241514

 View more versions of this product

Catalog No. AAA1241514


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Difluorophenyl isocyanate
1.31
50°C (122°F)
1.492
25 g
2831577
2,4-difluorophenyl isocyanate, 2,4-difluorophenylisocyanate, benzene, 2,4-difluoro-1-isocyanato, isocyanic acid 2,4-difluorophenyl ester, 2,4-difluoro-1-isocyanato-benzene, 2,4-difluoroisocyanatobenzene, timtec-bb sbb006537, pubchem4453, 2,4-diflurophenylisocyanate, acmc-1b06k
FC1=CC=C(N=C=O)C(F)=C1
155.10
155.11
59025-55-7
64°C to 66°C (10 mmHg)
C7H3F2NO
MFCD00001997
UN3080
Moisture sensitive
HNENEALJPWJWJY-UHFFFAOYSA-N
2,4-difluoro-1-isocyanatobenzene
593258
≥98%

Chemical Identifiers

59025-55-7
155.10
HNENEALJPWJWJY-UHFFFAOYSA-N
593258
FC1=CC=C(N=C=O)C(F)=C1
C7H3F2NO
MFCD00001997
2,4-difluorophenyl isocyanate, 2,4-difluorophenylisocyanate, benzene, 2,4-difluoro-1-isocyanato, isocyanic acid 2,4-difluorophenyl ester, 2,4-difluoro-1-isocyanato-benzene, 2,4-difluoroisocyanatobenzene, timtec-bb sbb006537, pubchem4453, 2,4-diflurophenylisocyanate, acmc-1b06k
2,4-difluoro-1-isocyanatobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H334-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 3; Packaging Group: II

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only