missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoromandelic acid, 97%

Catalog No. AAL1070803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1070803 1 g
AAL1070806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1070803 Supplier Thermo Scientific Chemicals Supplier No. L1070803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 132741-30-1 | C8H6F2O3 | 188.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 132741-30-1
Molecular Formula C8H6F2O3
Molecular Weight (g/mol) 188.13
MDL Number MFCD00061296
InChI Key RRRQFGNNRJHLNV-UHFFFAOYSA-N
Synonym 2,4-difluoromandelic acid, 2-2,4-difluorophenyl-2-hydroxyacetic acid, 2,4-difluorophenyl hydroxy acetic acid, 2,4-difluoro mandelic acid, 2,4-difluoro-a-hydroxyphenylacetic acid, alpha-hydroxy-2,4-difluorophenylacetic acid, 2,4-difluorophenyl hydroxy acetic acid #, 2,4-difluoro-alpha-hydroxybenzeneacetic acid, currency-hydroxy-2,4-difluorophenylacetic acid
PubChem CID 588651
IUPAC Name 2-(2,4-difluorophenyl)-2-hydroxyacetic acid
SMILES C1=CC(=C(C=C1F)F)C(C(=O)O)O

Specifications

Melting Point 122°C to 124°C
Quantity 1 g
Formula Weight 188.13
Percent Purity 97%
Chemical Name or Material 2,4-Difluoromandelic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only