Learn More
2,4-Difluorobenzoyl chloride, 97%
CAS: 72482-64-5 | C7H3ClF2O | 176.55 g/mol
$66.03 - $210.07
Chemical Identifiers
| CAS | 72482-64-5 |
|---|---|
| Molecular Formula | C7H3ClF2O |
| Molecular Weight (g/mol) | 176.55 |
| MDL Number | MFCD00000658 |
| InChI Key | JSWRVDNTKPAJLB-UHFFFAOYSA-N |
| PubChem CID | 588081 |
| IUPAC Name | 2,4-difluorobenzoyl chloride |
| SMILES | FC1=CC=C(C(Cl)=O)C(F)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1792006
|
Thermo Scientific Chemicals
A1792006 |
5 g |
Each for $66.03
|
|
|||||
|
AAA1792014
|
Thermo Scientific Chemicals
A1792014 |
25 g |
Each for $210.07
|
|
|||||
Description
2,4-Difluorobenzoyl chloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4-Difluorobenzoyl chloride is used as a pharmaceutical intermediate.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from water/ moisture, oxidizing agents, bases.
Chemical Identifiers
| 72482-64-5 | |
| 176.55 | |
| JSWRVDNTKPAJLB-UHFFFAOYSA-N | |
| 2,4-difluorobenzoyl chloride |
| C7H3ClF2O | |
| MFCD00000658 | |
| 588081 | |
| FC1=CC=C(C(Cl)=O)C(F)=C1 |
Specifications
| 72482-64-5 | |
| 96°C to 98°C (15 mmHg) | |
| C7H3ClF2O | |
| MFCD00000658 | |
| UN3265 | |
| Moisture sensitive | |
| Reacts with water. | |
| FC1=CC=C(C(Cl)=O)C(F)=C1 | |
| 176.55 | |
| 176.55 | |
| 2,4-Difluorobenzoyl chloride |
| 1.437 | |
| 82°C (179°F) | |
| 1.5145 | |
| 5 g | |
| 4800566 | |
| 2,4-difluorobenzoylchloride, benzoyl chloride, 2,4-difluoro, benzoyl chloride,2,4-difluoro, acmc-1bfet, 2,4difluorobenzoylchloride, 2,4-difluorobezoyl chloride, 2,4 difluorobenzoyl chloride, 2,4-difluoro benzoylchloride, 2,4-difluoro-benzoylchloride, 2,4-difluoro-benzoyl chloride | |
| JSWRVDNTKPAJLB-UHFFFAOYSA-N | |
| 2,4-difluorobenzoyl chloride | |
| 588081 | |
| 98% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 276-690-4
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only