missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Difluorobenzophenone 96.0+%, TCI America™

Catalog No. D17965G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17965G 5 g
D179625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17965G Supplier TCI America Supplier No. D17965G
Only null left

Chemical Identifiers

CAS 342-25-6
Molecular Formula C13H8F2O
Molecular Weight (g/mol) 218.203
MDL Number MFCD00000319
InChI Key LKFIWRPOVFNPKR-UHFFFAOYSA-N
Synonym 2,4'-difluorobenzophenone, 2-fluorophenyl 4-fluorophenyl methanone, 2',4-difluorobenzophenone, 2-fluorophenyl-4-fluorophenyl methanone, di2-fluorophenyl ketone, fr bvr df, pubchem7479, acmc-1clee, 2,4;-difluorobenzophenone, 2,4/'-difluorobenzophenone
PubChem CID 67651
IUPAC Name (2-fluorophenyl)-(4-fluorophenyl)methanone
SMILES C1=CC=C(C(=C1)C(=O)C2=CC=C(C=C2)F)F

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 218.20
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4′-Difluorobenzophenone