accessibility menu, dialog, popup

UserName

FOF

CAS RN 342-25-6

IUPAC Name: (2-fluorophenyl)(4-fluorophenyl)methanone
Synonyms: (2-fluorophenyl)(4-fluorophenyl)methanone
Molecular Weight (g/mol): 218.2
Molecular Formula: C13H8F2O
InChi Key: LKFIWRPOVFNPKR-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)C1=CC=CC=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results