missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzonitrile, 98%

CAS: 3939-09-1 | C7H3F2N | 139.11 g/mol

Supplier:  Thermo Scientific Chemicals A1411306

 View more versions of this product

Catalog No. AAA1411306


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Difluorobenzonitrile
43°C to 48°C
>110°C (230°F)
C7H3F2N
5 g
1940316
LJFDXXUKKMEQKE-UHFFFAOYSA-N
2,4-difluorobenzonitrile
77545
98%
3939-09-1
189°C
Almond-like
MFCD00009826
UN3439
2,4-difluorobenzenecarbonitrile, benzonitrile, 2,4-difluoro, 2,4-difluoro-benzonitrile, pubchem1563, acmc-1bohp, 2.4-difluoro benzonitrile, 2,4-difluoro benzonitrile, ksc493m5d, 2,4-difluorobenzonitrile, ljfdxxukkmeqke-uhfffaoysa
C1=CC(=C(C=C1F)F)C#N
139.11
139.11

Chemical Identifiers

3939-09-1
139.11
LJFDXXUKKMEQKE-UHFFFAOYSA-N
77545
C1=CC(=C(C=C1F)F)C#N
C7H3F2N
MFCD00009826
2,4-difluorobenzenecarbonitrile, benzonitrile, 2,4-difluoro, 2,4-difluoro-benzonitrile, pubchem1563, acmc-1bohp, 2.4-difluoro benzonitrile, 2,4-difluoro benzonitrile, ksc493m5d, 2,4-difluorobenzonitrile, ljfdxxukkmeqke-uhfffaoysa
2,4-difluorobenzonitrile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

EINECSNumber : 223-523-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only