missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Difluorobenzoic Acid 98.0+%, TCI America™
$72.70 - $700.89
Chemical Identifiers
| CAS | 1583-58-0 |
|---|---|
| Molecular Formula | C7H4F2O2 |
| Molecular Weight (g/mol) | 158.10 |
| MDL Number | MFCD00011670 |
| InChI Key | NJYBIFYEWYWYAN-UHFFFAOYSA-N |
| Synonym | 2,4-dfba, benzoic acid, 2,4-difluoro, 2,4-difluorobenzoicacid, 2,4-difluoro-benzoic acid, zlchem 472, pubchem1336, acmc-1brgf, 2,4-difluorbenzoic acid, 2,4-difluorobezoic acid, 2 4-difluorobenzoic acid |
| PubChem CID | 74102 |
| IUPAC Name | 2,4-difluorobenzoic acid |
| SMILES | OC(=O)C1=CC=C(F)C=C1F |
Chemical Identifiers
| 1583-58-0 | |
| 158.10 | |
| NJYBIFYEWYWYAN-UHFFFAOYSA-N | |
| 74102 | |
| OC(=O)C1=CC=C(F)C=C1F |
| C7H4F2O2 | |
| MFCD00011670 | |
| 2,4-dfba, benzoic acid, 2,4-difluoro, 2,4-difluorobenzoicacid, 2,4-difluoro-benzoic acid, zlchem 472, pubchem1336, acmc-1brgf, 2,4-difluorbenzoic acid, 2,4-difluorobezoic acid, 2 4-difluorobenzoic acid | |
| 2,4-difluorobenzoic acid |
Specifications
| 1583-58-0 | |
| C7H4F2O2 | |
| 25 g | |
| NJYBIFYEWYWYAN-UHFFFAOYSA-N | |
| 2,4-difluorobenzoic acid | |
| 74102 | |
| ≥98.0% (T) | |
| 2,4-Difluorobenzoic Acid |
| 188°C | |
| MFCD00011670 | |
| 2,4-dfba, benzoic acid, 2,4-difluoro, 2,4-difluorobenzoicacid, 2,4-difluoro-benzoic acid, zlchem 472, pubchem1336, acmc-1brgf, 2,4-difluorbenzoic acid, 2,4-difluorobezoic acid, 2 4-difluorobenzoic acid | |
| OC(=O)C1=CC=C(F)C=C1F | |
| 158.10 | |
| 158.10 | |
| Crystalline Powder |
Safety and Handling
TSCA : No