missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Difluorobenzaldehyde 98.0+%, TCI America™
$63.38 - $214.37
Chemical Identifiers
| CAS | 1550-35-2 |
|---|---|
| Molecular Formula | C7H4F2O |
| Molecular Weight (g/mol) | 142.11 |
| MDL Number | MFCD00010326 |
| InChI Key | WCGPCBACLBHDCI-UHFFFAOYSA-N |
| Synonym | 2,4-difluoro-benzaldehyde, benzaldehyde, 2,4-difluoro, pubchem2903, 2,4difluorobenzaldehyde, acmc-209dbv, 2,4-difluorbenzaldehyde, 2,4 difluorobenzaldehyde, 2.4-difluorobenzaldehyde, 2,4-difluoro benzaldehyde, 2.4-difluoro benzaldehyde |
| PubChem CID | 73770 |
| IUPAC Name | 2,4-difluorobenzaldehyde |
| SMILES | FC1=CC=C(C=O)C(F)=C1 |
Chemical Identifiers
| 1550-35-2 | |
| 142.11 | |
| WCGPCBACLBHDCI-UHFFFAOYSA-N | |
| 73770 | |
| FC1=CC=C(C=O)C(F)=C1 |
| C7H4F2O | |
| MFCD00010326 | |
| 2,4-difluoro-benzaldehyde, benzaldehyde, 2,4-difluoro, pubchem2903, 2,4difluorobenzaldehyde, acmc-209dbv, 2,4-difluorbenzaldehyde, 2,4 difluorobenzaldehyde, 2.4-difluorobenzaldehyde, 2,4-difluoro benzaldehyde, 2.4-difluoro benzaldehyde | |
| 2,4-difluorobenzaldehyde |
Specifications
| 1550-35-2 | |
| C7H4F2O | |
| 5 g | |
| 2,4-difluoro-benzaldehyde, benzaldehyde, 2,4-difluoro, pubchem2903, 2,4difluorobenzaldehyde, acmc-209dbv, 2,4-difluorbenzaldehyde, 2,4 difluorobenzaldehyde, 2.4-difluorobenzaldehyde, 2,4-difluoro benzaldehyde, 2.4-difluoro benzaldehyde | |
| FC1=CC=C(C=O)C(F)=C1 | |
| 142.11 | |
| 142.10 | |
| Liquid |
| Yellow | |
| MFCD00010326 | |
| 1989 | |
| WCGPCBACLBHDCI-UHFFFAOYSA-N | |
| 2,4-difluorobenzaldehyde | |
| 73770 | |
| ≥98.0% (GC) | |
| 2,4-Difluorobenzaldehyde |
Safety and Handling
TSCA : No