missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzaldehyde, 98%

Catalog No. AC271470050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC271470050 5 g Glass bottle
AC271470250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC271470050 Supplier Thermo Scientific Chemicals Supplier No. 271470050

CAS: 1550-35-2 | C7H4F2O | 142.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1550-35-2
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.11
MDL Number MFCD00010326
InChI Key WCGPCBACLBHDCI-UHFFFAOYSA-N
PubChem CID 73770
IUPAC Name 2,4-difluorobenzaldehyde
SMILES FC1=CC=C(C=O)C(F)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 2.0°C to 3.0°C
Color Colorless
Density 1.2900g/mL
Boiling Point 65.0°C to 66.0°C (17.0 mmHg)
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4975 to 1.4995 (20°C, 589nm)
Linear Formula F2C6H3CHO
Quantity 5 g
Specific Gravity 1.29
Synonym 2,4-difluoro-benzaldehyde, benzaldehyde, 2,4-difluoro, pubchem2903, 2,4difluorobenzaldehyde, acmc-209dbv, 2,4-difluorbenzaldehyde, 2,4 difluorobenzaldehyde, 2.4-difluorobenzaldehyde, 2,4-difluoro benzaldehyde, 2.4-difluoro benzaldehyde
Formula Weight 142.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 4-Difluorobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-287-2

RUO – Research Use Only