missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzaldehyde, 98%

CAS: 1550-35-2 | C7H4F2O | 142.11 g/mol

$110.86 - $333.64

Chemical Identifiers

CAS 1550-35-2
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.11
MDL Number MFCD00010326
InChI Key WCGPCBACLBHDCI-UHFFFAOYSA-N
PubChem CID 73770
IUPAC Name 2,4-difluorobenzaldehyde
SMILES FC1=CC=C(C=O)C(F)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1705206
View Documents
Thermo Scientific Chemicals
A1705206
5 g
Each for $110.86
Only null left
 
AAA1705214
View Documents
Thermo Scientific Chemicals
A1705214
25 g
Each for $333.64
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1550-35-2
142.11
WCGPCBACLBHDCI-UHFFFAOYSA-N
2,4-difluorobenzaldehyde
C7H4F2O
MFCD00010326
73770
FC1=CC=C(C=O)C(F)=C1

Specifications

1550-35-2
1.299
55°C (131°F)
1.498
5 g
2243422
2,4-difluoro-benzaldehyde, benzaldehyde, 2,4-difluoro, pubchem2903, 2,4difluorobenzaldehyde, acmc-209dbv, 2,4-difluorbenzaldehyde, 2,4 difluorobenzaldehyde, 2.4-difluorobenzaldehyde, 2,4-difluoro benzaldehyde, 2.4-difluoro benzaldehyde
FC1=CC=C(C=O)C(F)=C1
142.11
142.11
2,4-Difluorobenzaldehyde
2°C to 3°C
65°C to 66°C (17 mmHg)
C7H4F2O
MFCD00010326
UN1989
Air Sensitive
WCGPCBACLBHDCI-UHFFFAOYSA-N
2,4-difluorobenzaldehyde
73770
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.

EINECSNumber : 216-287-2

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only