missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzaldehyde, 98%

Catalog No. AAA1705206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1705206 5 g
AAA1705214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1705206 Supplier Thermo Scientific Chemicals Supplier No. A1705206

CAS: 1550-35-2 | C7H4F2O | 142.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1550-35-2
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.11
MDL Number MFCD00010326
InChI Key WCGPCBACLBHDCI-UHFFFAOYSA-N
PubChem CID 73770
IUPAC Name 2,4-difluorobenzaldehyde
SMILES FC1=CC=C(C=O)C(F)=C1

Specifications

Melting Point 2°C to 3°C
Density 1.299
Boiling Point 65°C to 66°C (17 mmHg)
Flash Point 55°C (131°F)
Refractive Index 1.498
Quantity 5 g
UN Number UN1989
Beilstein 2243422
Sensitivity Air Sensitive
Synonym 2,4-difluoro-benzaldehyde, benzaldehyde, 2,4-difluoro, pubchem2903, 2,4difluorobenzaldehyde, acmc-209dbv, 2,4-difluorbenzaldehyde, 2,4 difluorobenzaldehyde, 2.4-difluorobenzaldehyde, 2,4-difluoro benzaldehyde, 2.4-difluoro benzaldehyde
Formula Weight 142.11
Percent Purity 98%
Chemical Name or Material 2,4-Difluorobenzaldehyde
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber 216-287-2
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only