Learn More
2,4-Dichloropyrimidine, 98+%
CAS: 3934-20-1 | C4H2Cl2N2 | 148.974 g/mol
$102.03 - $2196.04
Chemical Identifiers
| CAS | 3934-20-1 |
|---|---|
| Molecular Formula | C4H2Cl2N2 |
| Molecular Weight (g/mol) | 148.974 |
| MDL Number | MFCD00006061 |
| InChI Key | BTTNYQZNBZNDOR-UHFFFAOYSA-N |
| PubChem CID | 77531 |
| IUPAC Name | 2,4-dichloropyrimidine |
| SMILES | C1=CN=C(N=C1Cl)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1513106
|
Thermo Scientific Chemicals
A1513106 |
5 g |
Each for $102.03
|
|
|||||
|
AAA1513109
|
Thermo Scientific Chemicals
A1513109 |
10 g |
Each for $141.22
|
|
|||||
|
AAA1513118
|
Thermo Scientific Chemicals
A1513118 |
50 g |
Each for $556.07
|
|
|||||
|
AAA1513130
|
Thermo Scientific Chemicals
A1513130 |
250 g |
Each for $2,196.04
|
|
|||||
Description
2,4-Dichloropyrimidine is used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4-Dichloropyrimidine is used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.
Solubility
Soluble in water (partly), methanol, chloroform, and ethyl acetate.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 3934-20-1 | |
| 148.974 | |
| BTTNYQZNBZNDOR-UHFFFAOYSA-N | |
| 2,4-dichloropyrimidine |
| C4H2Cl2N2 | |
| MFCD00006061 | |
| 77531 | |
| C1=CN=C(N=C1Cl)Cl |
Specifications
| 3934-20-1 | |
| 101°C (23 mmHg) | |
| MFCD00006061 | |
| 110911 | |
| pyrimidine, 2,4-dichloro, 2,6-dichloropyrimidine, 2,4-dichloro-pyrimidine, 2,4-dichlorpyrimidin, 2,4-dichloro pyrimidine, pyrimidine,2,4-dichloro, pyrimidine,4-dichloro, hydrochloride, zlchem 210, 2,4dichloropyrimidine | |
| BTTNYQZNBZNDOR-UHFFFAOYSA-N | |
| 2,4-dichloropyrimidine | |
| 77531 | |
| ≥98% |
| 57°C to 62°C | |
| C4H2Cl2N2 | |
| 5 g | |
| Moisture sensitive | |
| Soluble in water (partly),methanol,chloroform,and ethyl acetate. | |
| C1=CN=C(N=C1Cl)Cl | |
| 148.974 | |
| 148.98 | |
| 2,4-Dichloropyrimidine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 223-508-6
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only