missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloropyrimidine, 98+%

CAS: 3934-20-1 | C4H2Cl2N2 | 148.974 g/mol

Supplier:  Thermo Scientific Chemicals A1513118

 View more versions of this product

Catalog No. AAA1513118


Only null left
Explore available promotions

Description

Description

2,4-Dichloropyrimidine is used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichloropyrimidine is used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.

Solubility
Soluble in water (partly), methanol, chloroform, and ethyl acetate.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2,4-Dichloropyrimidine
57°C to 62°C
C4H2Cl2N2
50 g
Moisture sensitive
Soluble in water (partly),methanol,chloroform,and ethyl acetate.
C1=CN=C(N=C1Cl)Cl
148.974
148.98
3934-20-1
101°C (23 mmHg)
MFCD00006061
110911
pyrimidine, 2,4-dichloro, 2,6-dichloropyrimidine, 2,4-dichloro-pyrimidine, 2,4-dichlorpyrimidin, 2,4-dichloro pyrimidine, pyrimidine,2,4-dichloro, pyrimidine,4-dichloro, hydrochloride, zlchem 210, 2,4dichloropyrimidine
BTTNYQZNBZNDOR-UHFFFAOYSA-N
2,4-dichloropyrimidine
77531
≥98%

Chemical Identifiers

3934-20-1
148.974
BTTNYQZNBZNDOR-UHFFFAOYSA-N
2,4-dichloropyrimidine
C4H2Cl2N2
MFCD00006061
77531
C1=CN=C(N=C1Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 223-508-6

TSCA : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only