Learn More
2,4-Dichlorophenylhydrazine hydrochloride, 98%, Thermo Scientific™
$140.70 - $1917.12
Chemical Identifiers
| CAS | 5446-18-4 |
|---|---|
| Molecular Formula | C6H7Cl3N2 |
| Molecular Weight (g/mol) | 213.486 |
| MDL Number | MFCD00012929 |
| InChI Key | DDWYGJVFURAIJZ-UHFFFAOYSA-N |
| Synonym | 2,4-dichlorophenylhydrazine hydrochloride, 2,4-dichlorophenyl hydrazine hydrochloride, 2,4-dichlorophenyl hydrazine monohydrochloride, 2,4-dichlorophenylhydrazine hcl, 2,4-dichloro-phenyl-hydrazine hydrochloride, 1-2,4-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,4-dichlorophenyl-, monohydrochloride, pubchem7524, acmc-209lgs, ksc491e2h |
| PubChem CID | 2723905 |
| IUPAC Name | (2,4-dichlorophenyl)hydrazine;hydrochloride |
| SMILES | C1=CC(=C(C=C1Cl)Cl)NN.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1094309
|
Thermo Scientific Chemicals
A1094309 |
10 g |
Each for $140.70
|
|
|||||
|
AAA1094318
|
Thermo Scientific Chemicals
A1094318 |
50 g |
Each for $450.15
|
|
|||||
|
AAA1094330
|
Thermo Scientific Chemicals
A1094330 |
250 g |
Each for $1,917.12
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 5446-18-4 | |
| 213.486 | |
| DDWYGJVFURAIJZ-UHFFFAOYSA-N | |
| 2723905 | |
| C1=CC(=C(C=C1Cl)Cl)NN.Cl |
| C6H7Cl3N2 | |
| MFCD00012929 | |
| 2,4-dichlorophenylhydrazine hydrochloride, 2,4-dichlorophenyl hydrazine hydrochloride, 2,4-dichlorophenyl hydrazine monohydrochloride, 2,4-dichlorophenylhydrazine hcl, 2,4-dichloro-phenyl-hydrazine hydrochloride, 1-2,4-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,4-dichlorophenyl-, monohydrochloride, pubchem7524, acmc-209lgs, ksc491e2h | |
| (2,4-dichlorophenyl)hydrazine;hydrochloride |
Specifications
| 5446-18-4 | |
| C6H7Cl3N2 | |
| 10 g | |
| 2,4-dichlorophenylhydrazine hydrochloride, 2,4-dichlorophenyl hydrazine hydrochloride, 2,4-dichlorophenyl hydrazine monohydrochloride, 2,4-dichlorophenylhydrazine hcl, 2,4-dichloro-phenyl-hydrazine hydrochloride, 1-2,4-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,4-dichlorophenyl-, monohydrochloride, pubchem7524, acmc-209lgs, ksc491e2h | |
| C1=CC(=C(C=C1Cl)Cl)NN.Cl | |
| 213.486 | |
| 213.49 | |
| 2,4-Dichlorophenylhydrazine hydrochloride |
| ∼212°C (decomposition) | |
| MFCD00012929 | |
| 3708828 | |
| DDWYGJVFURAIJZ-UHFFFAOYSA-N | |
| (2,4-dichlorophenyl)hydrazine;hydrochloride | |
| 2723905 | |
| 98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 226-659-6
TSCA : No
Recommended Storage : Ambient temperatures