accessibility menu, dialog, popup

UserName

HClH2NNHClCl

CAS RN 5446-18-4

IUPAC Name: hydrogen (2,4-dichlorophenyl)hydrazine chloride
Synonyms: hydrogen (2,4-dichlorophenyl)hydrazine chloride
Molecular Weight (g/mol): 213.49
Molecular Formula: C6H7Cl3N2
InChi Key: DDWYGJVFURAIJZ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=C(Cl)C=C1Cl
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)