missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenylhydrazine hydrochloride, 98%, Thermo Scientific™

Catalog No. AAA1094309
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1094309 10 g
AAA1094318 50 g
AAA1094330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1094309 Supplier Thermo Scientific Chemicals Supplier No. A1094309

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 5446-18-4
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
MDL Number MFCD00012929
InChI Key DDWYGJVFURAIJZ-UHFFFAOYSA-N
Synonym 2,4-dichlorophenylhydrazine hydrochloride, 2,4-dichlorophenyl hydrazine hydrochloride, 2,4-dichlorophenyl hydrazine monohydrochloride, 2,4-dichlorophenylhydrazine hcl, 2,4-dichloro-phenyl-hydrazine hydrochloride, 1-2,4-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,4-dichlorophenyl-, monohydrochloride, pubchem7524, acmc-209lgs, ksc491e2h
PubChem CID 2723905
IUPAC Name (2,4-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=C(C=C1Cl)Cl)NN.Cl

Specifications

Melting Point ∼212°C (decomposition)
Quantity 10 g
Beilstein 3708828
Formula Weight 213.49
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorophenylhydrazine hydrochloride
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 226-659-6
TSCA No
Recommended Storage Ambient temperatures