missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7127296
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24335G 5 g
D243325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24335G Supplier TCI America Supplier No. D24335G
Only null left

Chemical Identifiers

CAS 5446-18-4
Molecular Formula C6H7Cl3N2
Molecular Weight (g/mol) 213.486
MDL Number MFCD00012929
InChI Key DDWYGJVFURAIJZ-UHFFFAOYSA-N
Synonym 2,4-dichlorophenylhydrazine hydrochloride, 2,4-dichlorophenyl hydrazine hydrochloride, 2,4-dichlorophenyl hydrazine monohydrochloride, 2,4-dichlorophenylhydrazine hcl, 2,4-dichloro-phenyl-hydrazine hydrochloride, 1-2,4-dichlorophenyl hydrazine hydrochloride, hydrazine, 2,4-dichlorophenyl-, monohydrochloride, pubchem7524, acmc-209lgs, ksc491e2h
PubChem CID 2723905
IUPAC Name (2,4-dichlorophenyl)hydrazine;hydrochloride
SMILES C1=CC(=C(C=C1Cl)Cl)NN.Cl

Specifications

Quantity 5 g
Formula Weight 213.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorophenylhydrazine Hydrochloride
EINECSNumber (3)-0472
TSCA No