missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichlorophenol 98.0+%, TCI America™
$75.08 - $704.61
Chemical Identifiers
| CAS | 120-83-2 |
|---|---|
| Molecular Formula | C6H4Cl2O |
| Molecular Weight (g/mol) | 163.00 |
| MDL Number | MFCD00002169 |
| InChI Key | HFZWRUODUSTPEG-UHFFFAOYSA-N |
| Synonym | 4,6-dichlorophenol, phenol, 2,4-dichloro, 2,4-dcp, 2,4-dichloro-phenol, 1-hydroxy-2,4-dichlorobenzene, rcra waste number u081, 2,4-dichlorohydroxybenzene, ccris 657, rcra waste no. u081, 1,3-dichloro-4-hydroxybenzene |
| PubChem CID | 8449 |
| ChEBI | CHEBI:16738 |
| IUPAC Name | 2,4-dichlorophenol |
| SMILES | OC1=CC=C(Cl)C=C1Cl |
Chemical Identifiers
| 120-83-2 | |
| 163.00 | |
| HFZWRUODUSTPEG-UHFFFAOYSA-N | |
| 8449 | |
| 2,4-dichlorophenol |
| C6H4Cl2O | |
| MFCD00002169 | |
| 4,6-dichlorophenol, phenol, 2,4-dichloro, 2,4-dcp, 2,4-dichloro-phenol, 1-hydroxy-2,4-dichlorobenzene, rcra waste number u081, 2,4-dichlorohydroxybenzene, ccris 657, rcra waste no. u081, 1,3-dichloro-4-hydroxybenzene | |
| CHEBI:16738 | |
| OC1=CC=C(Cl)C=C1Cl |
Specifications
| 120-83-2 | |
| White-Yellow | |
| C6H4Cl2O | |
| 25 g | |
| 4,6-dichlorophenol, phenol, 2,4-dichloro, 2,4-dcp, 2,4-dichloro-phenol, 1-hydroxy-2,4-dichlorobenzene, rcra waste number u081, 2,4-dichlorohydroxybenzene, ccris 657, rcra waste no. u081, 1,3-dichloro-4-hydroxybenzene | |
| OC1=CC=C(Cl)C=C1Cl | |
| 163.00 | |
| CHEBI:16738 | |
| ≥98.0% (GC) | |
| 2,4-Dichlorophenol |
| 42°C | |
| 210°C | |
| MFCD00002169 | |
| 2020 | |
| HFZWRUODUSTPEG-UHFFFAOYSA-N | |
| 2,4-dichlorophenol | |
| 8449 | |
| 163.00 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-0903&(3)-0930
RTECSNumber : SK8575000
TSCA : Yes