missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenol 98.0+%, TCI America™

Catalog No. p-7127961
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
D38651G 1 g
D38655G 5 g
2 options

Catalog No. D38651G

Supplier: TCI America D38651G

Only null left

Chemical Identifiers

CAS 120-83-2
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002169
InChI Key HFZWRUODUSTPEG-UHFFFAOYSA-N
Synonym 4,6-dichlorophenol, phenol, 2,4-dichloro, 2,4-dcp, 2,4-dichloro-phenol, 1-hydroxy-2,4-dichlorobenzene, rcra waste number u081, 2,4-dichlorohydroxybenzene, ccris 657, rcra waste no. u081, 1,3-dichloro-4-hydroxybenzene
PubChem CID 8449
ChEBI CHEBI:16738
IUPAC Name 2,4-dichlorophenol
SMILES OC1=CC=C(Cl)C=C1Cl

Specifications

Melting Point 43°C
Color White
Boiling Point 210°C
Quantity 1 g
UN Number 2020
Formula Weight 163.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorophenol [for Biochemical Research]
EINECSNumber (3)-0903&(3)-0930
RTECSNumber SK8575000
TSCA Yes