missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloronitrobenzene 99.0+%, TCI America™
Supplier: TCI America D0982500G
Specifications
| 2,4-Dichloronitrobenzene | |
| White-Yellow | |
| C6H3Cl2NO2 | |
| 500 g | |
| 2,4-dichloronitrobenzene, 1,3-dichloro-4-nitrobenzene, benzene, 2,4-dichloro-1-nitro, unii-85p1a896rr, ccris 3095, benzene, 1,3-dichloro-4-nitro, nitro-m-dichlorobenzene, pubchem15890, 2,4-dichloronitrobenzen, 4-nitro-m-dichlorobenzene | |
| C1=CC(=C(C=C1Cl)Cl)[N+](=O)[O-] | |
| 191.995 | |
| 192.00 | |
| Crystalline Powder |
| 611-06-3 | |
| 259°C | |
| MFCD00007071 | |
| 2811 | |
| QUIMTLZDMCNYGY-UHFFFAOYSA-N | |
| 2,4-dichloro-1-nitrobenzene | |
| 11899 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 611-06-3 | |
| 191.995 | |
| QUIMTLZDMCNYGY-UHFFFAOYSA-N | |
| 11899 | |
| C1=CC(=C(C=C1Cl)Cl)[N+](=O)[O-] |
| C6H3Cl2NO2 | |
| MFCD00007071 | |
| 2,4-dichloronitrobenzene, 1,3-dichloro-4-nitrobenzene, benzene, 2,4-dichloro-1-nitro, unii-85p1a896rr, ccris 3095, benzene, 1,3-dichloro-4-nitro, nitro-m-dichlorobenzene, pubchem15890, 2,4-dichloronitrobenzen, 4-nitro-m-dichlorobenzene | |
| 2,4-dichloro-1-nitrobenzene |
Safety and Handling
EINECSNumber : (3)-0455
RTECSNumber : CZ5420000
TSCA : Yes