missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloronitrobenzene 99.0+%, TCI America™

Catalog No. p-7126538
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D098225G 25 g
D0982500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D098225G Supplier TCI America Supplier No. D098225G
Only null left

Chemical Identifiers

CAS 611-06-3
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 191.995
MDL Number MFCD00007071
InChI Key QUIMTLZDMCNYGY-UHFFFAOYSA-N
Synonym 2,4-dichloronitrobenzene, 1,3-dichloro-4-nitrobenzene, benzene, 2,4-dichloro-1-nitro, unii-85p1a896rr, ccris 3095, benzene, 1,3-dichloro-4-nitro, nitro-m-dichlorobenzene, pubchem15890, 2,4-dichloronitrobenzen, 4-nitro-m-dichlorobenzene
PubChem CID 11899
IUPAC Name 2,4-dichloro-1-nitrobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)[N+](=O)[O-]

Specifications

Color White-Yellow
Boiling Point 259°C
Quantity 25 g
UN Number 2811
Formula Weight 192.00
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloronitrobenzene
EINECSNumber (3)-0455
RTECSNumber CZ5420000
TSCA Yes