missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzylamine, 98%

Catalog No. p-7021039
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1320706 5 g
AAA1320714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1320706 Supplier Thermo Scientific Chemicals Supplier No. A1320706
Only null left

CAS: 95-00-1 | C7H7Cl2N | 176.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-00-1
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00008109
InChI Key SJUKJZSTBBSGHF-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzylamine, 2,4-dichlorophenyl methanamine, benzenemethanamine, 2,4-dichloro, benzylamine, 2,4-dichloro, 2,4-dichlorobenzyl amine, 1-2,4-dichlorophenyl methanamine, 2,4-dichloro-benzylamine, 2,4-dichlorophenyl methylamine, o,p-dichlorobenzylamine, acmc-209rur
PubChem CID 1485
IUPAC Name (2,4-dichlorophenyl)methanamine
SMILES NCC1=CC=C(Cl)C=C1Cl

Specifications

Density 1.32
Boiling Point 258°C to 260°C
Flash Point >110°C (230°F)
Odor Amine-like
Refractive Index 1.578
Quantity 5 g
UN Number UN2735
Beilstein 775667
Sensitivity Air Sensitive
Formula Weight 176.05
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorobenzylamine
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 202-382-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only