missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzyl Alcohol 97.0+%, TCI America™

Catalog No. D183725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D183725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D183725G Supplier TCI America Supplier No. D183725G
Only null left

Chemical Identifiers

CAS 1777-82-8
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00004606
InChI Key DBHODFSFBXJZNY-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzyl alcohol, 2,4-dichlorophenyl methanol, dybenal, 2,4-dichlorobenzenemethanol, rapidosept, myacide sp, benzenemethanol, 2,4-dichloro, benzyl alcohol, 2,4-dichloro, unii-1nkx3648j9, 2,4-dichlorobenzylalcohol
PubChem CID 15684
ChEBI CHEBI:48220
IUPAC Name (2,4-dichlorophenyl)methanol
SMILES C1=CC(=C(C=C1Cl)Cl)CO

Specifications

Melting Point 59°C
Color White-Yellow
Boiling Point 150°C
Quantity 25 g
Formula Weight 177.02
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorobenzyl Alcohol
RTECSNumber DO0890000
TSCA No