accessibility menu, dialog, popup

UserName

HOClCl

CAS RN 1777-82-8

IUPAC Name: (2,4-dichlorophenyl)methanol
Synonyms: 2,4-dichlorobenzyl alcohol myacide SP C7H6cl2O rapidosept dichlorobenzyl alcohol dybenal
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: DBHODFSFBXJZNY-UHFFFAOYSA-N
SMILES: OCC1=CC=C(Cl)C=C1Cl
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)