missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichlorobenzyl alcohol, 99%
Description
2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.
Solubility
Soluble in water, methanol and chloroform.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.
Solubility
Soluble in water, methanol and chloroform.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 57°C to 61°C |
| Flash Point | >110°C (230°F) |
| Quantity | 25 g |
| Beilstein | 1448652 |
| Merck Index | 14,3060 |
| Solubility Information | Soluble in water,methanol and chloroform. |
| Formula Weight | 177.03 |
| Percent Purity | 99% |
| Chemical Name or Material | 2,4-Dichlorobenzyl alcohol |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement H318 Causes serious eye damage. |
| Precautionary Statement | GHS P Statement P280i-P305+P351+P338-P337+P313 Wear eye/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF eye irritation persists: Get medical advice/attention. |
| EINECSNumber | 217-210-5 |
| RTECSNumber | DO0890000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only