missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzyl alcohol, 99%

Catalog No. AAA1333814
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1333814 25 g
AAA1333822 100 g
AAA1333836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1333814 Supplier Thermo Scientific Chemicals Supplier No. A1333814
Only null left

CAS: 1777-82-8 | C7H6Cl2O | 177.024 g/mol

2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichlorobenzyl alcohol is an antiseptic found in commercially available throat lozenges. Also used deactivate Respiratory syncytial virus and SARS-Cov.

Solubility
Soluble in water, methanol and chloroform.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 1777-82-8
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00004606
InChI Key DBHODFSFBXJZNY-UHFFFAOYSA-N
Synonym 2,4-dichlorobenzyl alcohol, 2,4-dichlorophenyl methanol, dybenal, 2,4-dichlorobenzenemethanol, rapidosept, myacide sp, benzenemethanol, 2,4-dichloro, benzyl alcohol, 2,4-dichloro, unii-1nkx3648j9, 2,4-dichlorobenzylalcohol
PubChem CID 15684
ChEBI CHEBI:48220
IUPAC Name (2,4-dichlorophenyl)methanol
SMILES C1=CC(=C(C=C1Cl)Cl)CO

Specifications

Melting Point 57°C to 61°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1448652
Merck Index 14,3060
Solubility Information Soluble in water,methanol and chloroform.
Formula Weight 177.03
Percent Purity 99%
Chemical Name or Material 2,4-Dichlorobenzyl alcohol
Hazard Category Danger
Hazard Statement GHS H Statement
H318
Causes serious eye damage.
Precautionary Statement GHS P Statement
P280i-P305+P351+P338-P337+P313
Wear eye/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
EINECSNumber 217-210-5
RTECSNumber DO0890000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only