missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichlorobenzenethiol 97.0+%, TCI America™
Supplier: TCI America D1938250G
Specifications
| 2,4-Dichlorobenzenethiol | |
| Yellow | |
| C6H4Cl2S | |
| 250 g | |
| FGBVJFREPSJSNG-UHFFFAOYSA-N | |
| 2,4-dichlorobenzenethiol | |
| 70730 | |
| ≥97.0% (GC) |
| 1122-41-4 | |
| 115°C | |
| MFCD00041421 | |
| 2,4-dichlorothiophenol, 2,4-dichlorobenzene-1-thiol, benzenethiol, 2,4-dichloro, 2,4-dichloro thiophenol, pubchem6798, 2,4 dichlorothiophenol, acmc-1buvb, 2,4-dichlorothio-phenol, 2,4-dichloro-benzenethiol, # | |
| C1=CC(=C(C=C1Cl)Cl)S | |
| 179.058 | |
| 179.06 | |
| Liquid |
Chemical Identifiers
| 1122-41-4 | |
| 179.058 | |
| FGBVJFREPSJSNG-UHFFFAOYSA-N | |
| 70730 | |
| C1=CC(=C(C=C1Cl)Cl)S |
| C6H4Cl2S | |
| MFCD00041421 | |
| 2,4-dichlorothiophenol, 2,4-dichlorobenzene-1-thiol, benzenethiol, 2,4-dichloro, 2,4-dichloro thiophenol, pubchem6798, 2,4 dichlorothiophenol, acmc-1buvb, 2,4-dichlorothio-phenol, 2,4-dichloro-benzenethiol, # | |
| 2,4-dichlorobenzenethiol |
Safety and Handling
TSCA : No