missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzenethiol 97.0+%, TCI America™

Catalog No. p-7127024
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D193825G 25 g
D1938250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D193825G Supplier TCI America Supplier No. D193825G
Only null left

Chemical Identifiers

CAS 1122-41-4
Molecular Formula C6H4Cl2S
Molecular Weight (g/mol) 179.058
MDL Number MFCD00041421
InChI Key FGBVJFREPSJSNG-UHFFFAOYSA-N
Synonym 2,4-dichlorothiophenol, 2,4-dichlorobenzene-1-thiol, benzenethiol, 2,4-dichloro, 2,4-dichloro thiophenol, pubchem6798, 2,4 dichlorothiophenol, acmc-1buvb, 2,4-dichlorothio-phenol, 2,4-dichloro-benzenethiol, #
PubChem CID 70730
IUPAC Name 2,4-dichlorobenzenethiol
SMILES C1=CC(=C(C=C1Cl)Cl)S

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
Formula Weight 179.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dichlorobenzenethiol