missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzamide 98.0+%, TCI America™

Catalog No. D286725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D286725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D286725G Supplier TCI America Supplier No. D286725G
Only null left

Chemical Identifiers

CAS 2447-79-2
Molecular Formula C7H5Cl2NO
Molecular Weight (g/mol) 190.023
MDL Number MFCD00007974
InChI Key XXFUNTSOBHSMBU-UHFFFAOYSA-N
Synonym benzamide, 2,4-dichloro, unii-zx274bu5ra, zx274bu5ra, 2,4-dichloro-benzamide, acmc-1crzq, xxfuntsobhsmbu-uhfffaoysa
PubChem CID 75556
IUPAC Name 2,4-dichlorobenzamide
SMILES C1=CC(=C(C=C1Cl)Cl)C(=O)N

Specifications

Melting Point 193°C
Quantity 25 g
Formula Weight 190.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorobenzamide