accessibility menu, dialog, popup

UserName

H2NOClCl

CAS RN 2447-79-2

IUPAC Name: 2,4-dichlorobenzamide
Synonyms: 2,4-dichlorobenzamide
Molecular Weight (g/mol): 190.02
Molecular Formula: C7H5Cl2NO
InChi Key: XXFUNTSOBHSMBU-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(Cl)C=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results