missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylphenol, 98%

Catalog No. p-7055672
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6431203 1 g
AAH6431206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6431203 Supplier Thermo Scientific Chemicals Supplier No. H6431203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1570-65-6 | C7H6Cl2O | 177.024 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1570-65-6
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00019988
InChI Key WJQZZLQMLJPKQH-UHFFFAOYSA-N
PubChem CID 15292
IUPAC Name 2,4-dichloro-6-methylphenol
SMILES CC1=CC(=CC(=C1O)Cl)Cl

Specifications

Melting Point 52°C to 56°C
Color Brown
Boiling Point 225°C (literature)
Quantity 1 g
UN Number UN3077
Beilstein 1864865
Synonym 4,6-dichloro-o-cresol, 4,6-dichloro-2-methylphenol, phenol, 2,4-dichloro-6-methyl, o-cresol, 4,6-dichloro, 4,6-dichlorocresol, phenol,2,4-dichloro-6-methyl, dichlorocresol, o-cresol,6-dichloro, 4,6-dichlor-o-kresol, acmc-1c0mk
Formula Weight 177.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,4-Dichloro-6-methylphenol
Show More Show Less
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 216-382-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only