missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-methylbenzylamine, 98+%, Thermo Scientific™

Catalog No. p-7046739
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Catalog No. Quantity
AAL1610803 1 g
1 options

Catalog No. AAL1610803

Supplier: Thermo Scientific Chemicals L1610803

Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 150517-76-3
Molecular Formula C8H9Cl2N
Molecular Weight (g/mol) 190.07
MDL Number MFCD00052913
InChI Key QISTWSMZJRTDCS-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-methylbenzylamine, 2,4-dichloro-6-methylphenyl methanamine, 2,4-dichloro-6-methylphenyl methylamine, benzenemethanamine,2,4-dichloro-6-methyl, benzenemethanamine, 2,4-dichloro-6-methyl, 2,4-dichloro-6-methylbenzyl amine hydrochloride, 4,6-dichloro-2-methylbenzylamine, acmc-1c3je, 2-aminomethyl-3,5-dichlorotoluene, 1-2,4-dichloro-6-methylphenyl methanamine
PubChem CID 2800971
SMILES CC1=CC(Cl)=CC(Cl)=C1CN

Specifications

Melting Point 52°C to 56°C
Quantity 1 g
UN Number UN3259
Beilstein 8615227
Sensitivity Air Sensitive
IUPAC Name (2,4-dichloro-6-methylphenyl)methanamine
Formula Weight 190.07
Percent Purity ≥98%
Chemical Name or Material 2,4-Dichloro-6-methylbenzylamine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures