accessibility menu, dialog, popup

UserName

CH3NH2ClCl

CAS RN 150517-76-3

IUPAC Name: 1-(2,4-dichloro-6-methylphenyl)methanamine
Synonyms: 1-(2,4-dichloro-6-methylphenyl)methanamine
Molecular Weight (g/mol): 190.07
Molecular Formula: C8H9Cl2N
InChi Key: QISTWSMZJRTDCS-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC(Cl)=C1CN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results