missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-sulfamoylbenzoic Acid 97.0+%, TCI America™

Catalog No. D254125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D254125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D254125G Supplier TCI America Supplier No. D254125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2736-23-4
Molecular Formula C7H5Cl2NO4S
Molecular Weight (g/mol) 270.08
MDL Number MFCD00007931
InChI Key ZSHHRBYVHTVRFK-UHFFFAOYSA-N
Synonym lasamide, 5-aminosulfonyl-2,4-dichlorobenzoic acid, 2,4-dichloro-5-sulphamoylbenzoic acid, 2,4-dichloro-5-sulfamoyl-benzoic acid, unii-leg53tf0sn, 2,4-dichloro-5-sulfamylbenzoic acid, benzoic acid, 5-aminosulfonyl-2,4-dichloro, leg53tf0sn, 3-sulfamoyl-4,6-dichlorobenzoic acid, benzoic acid, 2,4-dichloro-5-sulfamoyl
PubChem CID 17655
IUPAC Name 2,4-dichloro-5-sulfamoylbenzoic acid
SMILES C1=C(C(=CC(=C1S(=O)(=O)N)Cl)Cl)C(=O)O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 270.08
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-5-sulfamoylbenzoic Acid
EINECSNumber (9)-0583
RTECSNumber DG8100000
TSCA No