accessibility menu, dialog, popup

UserName

CAS RN 2736-23-4

IUPAC Name: sodium 2,4-dichloro-5-sulfamoylbenzoate
Synonyms: sodium 2,4-dichloro-5-sulfamoylbenzoate
Molecular Weight (g/mol): 292.06
Molecular Formula: C7H4Cl2NNaO4S
InChi Key: NUMZLXREOJYJJN-UHFFFAOYSA-M
SMILES: [Na+].NS(=O)(=O)C1=CC(C([O-])=O)=C(Cl)C=C1Cl
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)