missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-sulfamoylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog No. p-7027660
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2458918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2458918 Supplier Thermo Scientific Chemicals Supplier No. B2458918

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2736-23-4
Molecular Formula C7H5Cl2NO4S
Molecular Weight (g/mol) 270.08
MDL Number MFCD00007931
InChI Key ZSHHRBYVHTVRFK-UHFFFAOYSA-N
Synonym lasamide, 5-aminosulfonyl-2,4-dichlorobenzoic acid, 2,4-dichloro-5-sulphamoylbenzoic acid, 2,4-dichloro-5-sulfamoyl-benzoic acid, unii-leg53tf0sn, 2,4-dichloro-5-sulfamylbenzoic acid, benzoic acid, 5-aminosulfonyl-2,4-dichloro, leg53tf0sn, 3-sulfamoyl-4,6-dichlorobenzoic acid, benzoic acid, 2,4-dichloro-5-sulfamoyl
PubChem CID 17655
IUPAC Name 2,4-dichloro-5-sulfamoylbenzoic acid
SMILES C1=C(C(=CC(=C1S(=O)(=O)N)Cl)Cl)C(=O)O

Specifications

Melting Point 230°C to 232°C (decomposition)
Quantity 50 g
Formula Weight 270.09
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-5-sulfamoylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-358-3
RTECSNumber DG8100000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only