missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloro-5-nitropyrimidine 98.0+%, TCI America™
$106.25 - $315.03
Chemical Identifiers
| CAS | 49845-33-2 |
|---|---|
| Molecular Formula | C4HCl2N3O2 |
| Molecular Weight (g/mol) | 193.97 |
| MDL Number | MFCD00127867 |
| InChI Key | INUSQTPGSHFGHM-UHFFFAOYSA-N |
| Synonym | 2,4-dichloro-5-nitro-pyrimidine, pyrimidine, 2,4-dichloro-5-nitro, 2, 4-dichloro-5-nitropyrimidine, 3-nitro-2,4-dichloropyrimidine, 2,4-dichloro-5-nitro pyrimidine, zlchem 540, pubchem7079, acmc-1ann4, ksc236i7h, 2,4-dichloro5-nitropyrimidine |
| PubChem CID | 521266 |
| IUPAC Name | 2,4-dichloro-5-nitropyrimidine |
| SMILES | [O-][N+](=O)C1=CN=C(Cl)N=C1Cl |
Chemical Identifiers
| 49845-33-2 | |
| 193.97 | |
| INUSQTPGSHFGHM-UHFFFAOYSA-N | |
| 521266 | |
| [O-][N+](=O)C1=CN=C(Cl)N=C1Cl |
| C4HCl2N3O2 | |
| MFCD00127867 | |
| 2,4-dichloro-5-nitro-pyrimidine, pyrimidine, 2,4-dichloro-5-nitro, 2, 4-dichloro-5-nitropyrimidine, 3-nitro-2,4-dichloropyrimidine, 2,4-dichloro-5-nitro pyrimidine, zlchem 540, pubchem7079, acmc-1ann4, ksc236i7h, 2,4-dichloro5-nitropyrimidine | |
| 2,4-dichloro-5-nitropyrimidine |
Specifications
| 49845-33-2 | |
| C4HCl2N3O2 | |
| 5 g | |
| INUSQTPGSHFGHM-UHFFFAOYSA-N | |
| 2,4-dichloro-5-nitropyrimidine | |
| 521266 | |
| ≥98.0% (GC) | |
| 2,4-Dichloro-5-nitropyrimidine |
| 139°C | |
| MFCD00127867 | |
| 2,4-dichloro-5-nitro-pyrimidine, pyrimidine, 2,4-dichloro-5-nitro, 2, 4-dichloro-5-nitropyrimidine, 3-nitro-2,4-dichloropyrimidine, 2,4-dichloro-5-nitro pyrimidine, zlchem 540, pubchem7079, acmc-1ann4, ksc236i7h, 2,4-dichloro5-nitropyrimidine | |
| [O-][N+](=O)C1=CN=C(Cl)N=C1Cl | |
| 193.97 | |
| 193.97 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator