missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-methylbenzenesulfonyl chloride, 97%

CAS: 28286-86-4 | C7H5Cl3O2S | 259.53 g/mol

Supplier:  Thermo Scientific Chemicals A1971809

 View more versions of this product

Catalog No. AAA1971809


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Dichloro-5-methylbenzenesulfonyl chloride
70°C to 73°C
MFCD00044016
UN3261
2,4-dichloro-5-methylbenzene-1-sulfonyl chloride, 2,4-dichloro-5-methylbenzenesulfonylchloride, 2,4-dichloro-5-methylbenzene-1-sulphonyl chloride, 2,4-dichloro-5-methylphenylsulfonyl chloride, acmc-1cpwo, 2,4-dichloro-5-methylphenyl chlorosulfone, 2,4-dichloro-5-methyl-benzenesulfonyl chloride, 2,4-dichloro-5-methylbenzene sulfonyl chloride, 2,4-dichloro-5-methylbenzenesulphonyl chloride, benzenesulfonylchloride,2,4-dichloro-5-methyl
CC1=CC(=C(Cl)C=C1Cl)S(Cl)(=O)=O
259.53
259.54
28286-86-4
C7H5Cl3O2S
10 g
Moisture sensitive
DFBKCDYLPLBSJP-UHFFFAOYSA-N
2,4-dichloro-5-methylbenzenesulfonyl chloride
119914
97%

Chemical Identifiers

28286-86-4
259.53
DFBKCDYLPLBSJP-UHFFFAOYSA-N
119914
C7H5Cl3O2S
MFCD00044016
2,4-dichloro-5-methylbenzene-1-sulfonyl chloride, 2,4-dichloro-5-methylbenzenesulfonylchloride, 2,4-dichloro-5-methylbenzene-1-sulphonyl chloride, 2,4-dichloro-5-methylphenylsulfonyl chloride, acmc-1cpwo, 2,4-dichloro-5-methylphenyl chlorosulfone, 2,4-dichloro-5-methyl-benzenesulfonyl chloride, 2,4-dichloro-5-methylbenzene sulfonyl chloride, 2,4-dichloro-5-methylbenzenesulphonyl chloride, benzenesulfonylchloride,2,4-dichloro-5-methyl
CC1=CC(=C(Cl)C=C1Cl)S(Cl)(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 248-938-1

RTECSNumber : QR8650000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only