missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-methylbenzenesulfonyl chloride, 97%

Catalog No. p-7024659
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1971888 2.5 g
AAA1971809 10 g
AAA1971818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1971888 Supplier Thermo Scientific Chemicals Supplier No. A1971888

CAS: 28286-86-4 | C7H5Cl3O2S | 259.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28286-86-4
Molecular Formula C7H5Cl3O2S
Molecular Weight (g/mol) 259.53
MDL Number MFCD00044016
InChI Key DFBKCDYLPLBSJP-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-methylbenzene-1-sulfonyl chloride, 2,4-dichloro-5-methylbenzenesulfonylchloride, 2,4-dichloro-5-methylbenzene-1-sulphonyl chloride, 2,4-dichloro-5-methylphenylsulfonyl chloride, acmc-1cpwo, 2,4-dichloro-5-methylphenyl chlorosulfone, 2,4-dichloro-5-methyl-benzenesulfonyl chloride, 2,4-dichloro-5-methylbenzene sulfonyl chloride, 2,4-dichloro-5-methylbenzenesulphonyl chloride, benzenesulfonylchloride,2,4-dichloro-5-methyl
PubChem CID 119914
SMILES CC1=CC(=C(Cl)C=C1Cl)S(Cl)(=O)=O

Specifications

Melting Point 70°C to 73°C
Quantity 2.5 g
UN Number UN3261
Sensitivity Moisture sensitive
IUPAC Name 2,4-dichloro-5-methylbenzenesulfonyl chloride
Formula Weight 259.54
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-5-methylbenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8
EINECSNumber 248-938-1
RTECSNumber QR8650000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only