accessibility menu, dialog, popup

UserName

CH3ClClSClOO

CAS RN 28286-86-4

IUPAC Name: 2,4-dichloro-5-methylbenzene-1-sulfonyl chloride
Synonyms: 2,4-dichloro-5-methylbenzenesulfonyl chloride
Molecular Weight (g/mol): 259.53
Molecular Formula: C7H5Cl3O2S
InChi Key: DFBKCDYLPLBSJP-UHFFFAOYSA-N
SMILES: CC1=CC(=C(Cl)C=C1Cl)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)