missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-isopropoxyaniline, 98%

CAS: 41200-96-8 | C9H11Cl2NO | 220.093 g/mol

Supplier:  Thermo Scientific Chemicals L1837106

 View more versions of this product

Catalog No. AAL1837106


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Dichloro-5-isopropoxyaniline
40°C to 44°C
C9H11Cl2NO
5 g
2836827
RWRWIICNMOKBIQ-UHFFFAOYSA-N
2,4-dichloro-5-propan-2-yloxyaniline
170476
98%
41200-96-8
110°C to 112°C (0.2 mmHg)
MFCD00974411
UN2811
2,4-dichloro-5-isopropoxyaniline, 2,4-dichloro-5-2-propyloxy aniline, 2,4-dichloro-5-propan-2-yloxy aniline, benzenamine, 2,4-dichloro-5-1-methylethoxy, acmc-20aodr, 2,4-dichloro-5-isopropyloxyaniline, 2,4-dichloro-5-isopropoxyphenylamine, 2,4-dichloro-5-isopropoxy-phenylamine, 2,4-dichloro-5-1-methylethoxy aniline, 2,4-dichloro-5-methylethoxy phenylamine
CC(C)OC1=C(C=C(C(=C1)N)Cl)Cl
220.093
220.11

Chemical Identifiers

41200-96-8
220.093
RWRWIICNMOKBIQ-UHFFFAOYSA-N
170476
CC(C)OC1=C(C=C(C(=C1)N)Cl)Cl
C9H11Cl2NO
MFCD00974411
2,4-dichloro-5-isopropoxyaniline, 2,4-dichloro-5-2-propyloxy aniline, 2,4-dichloro-5-propan-2-yloxy aniline, benzenamine, 2,4-dichloro-5-1-methylethoxy, acmc-20aodr, 2,4-dichloro-5-isopropyloxyaniline, 2,4-dichloro-5-isopropoxyphenylamine, 2,4-dichloro-5-isopropoxy-phenylamine, 2,4-dichloro-5-1-methylethoxy aniline, 2,4-dichloro-5-methylethoxy phenylamine
2,4-dichloro-5-propan-2-yloxyaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 255-258-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only