missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-isopropoxyaniline, 98%

Catalog No. p-7047260
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1837103 1 g
AAL1837106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1837103 Supplier Thermo Scientific Chemicals Supplier No. L1837103
Only null left

CAS: 41200-96-8 | C9H11Cl2NO | 220.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 41200-96-8
Molecular Formula C9H11Cl2NO
Molecular Weight (g/mol) 220.093
MDL Number MFCD00974411
InChI Key RWRWIICNMOKBIQ-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-isopropoxyaniline, 2,4-dichloro-5-2-propyloxy aniline, 2,4-dichloro-5-propan-2-yloxy aniline, benzenamine, 2,4-dichloro-5-1-methylethoxy, acmc-20aodr, 2,4-dichloro-5-isopropyloxyaniline, 2,4-dichloro-5-isopropoxyphenylamine, 2,4-dichloro-5-isopropoxy-phenylamine, 2,4-dichloro-5-1-methylethoxy aniline, 2,4-dichloro-5-methylethoxy phenylamine
PubChem CID 170476
IUPAC Name 2,4-dichloro-5-propan-2-yloxyaniline
SMILES CC(C)OC1=C(C=C(C(=C1)N)Cl)Cl

Specifications

Melting Point 40°C to 44°C
Boiling Point 110°C to 112°C (0.2 mmHg)
Quantity 1 g
UN Number UN2811
Beilstein 2836827
Formula Weight 220.11
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-5-isopropoxyaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 255-258-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only