missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-fluorobenzonitrile, 97%

Catalog No. AAH2646409
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2646409 10 g
AAH2646403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2646409 Supplier Thermo Scientific Chemicals Supplier No. H2646409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 128593-93-1 | C7H2Cl2FN | 190.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 128593-93-1
Molecular Formula C7H2Cl2FN
Molecular Weight (g/mol) 190.00
MDL Number MFCD03840469
InChI Key XZBZWBDTFUFZSU-UHFFFAOYSA-N
PubChem CID 2782753
IUPAC Name 2,4-dichloro-5-fluorobenzonitrile
SMILES FC1=CC(C#N)=C(Cl)C=C1Cl

Specifications

Melting Point 44°C to 47°C
Quantity 10 g
UN Number UN3439
Synonym 2,4-dichloro-5-fluoro-benzonitrile, pubchem1566, acmc-1bxxv, 2,4-dichloro-5-fluoro benzonitrile, benzonitrile,2,4-dichloro-5-fluoro, benzonitrile, 2,4-dichloro-5-fluoro, 2,4-dichloro-5-fluorobenzonitrile, 2,4-dichloro-5-fluorobenzenecarbonitrile, 2,4-bis chloranyl-5-fluoranyl-benzenecarbonitrile
Formula Weight 190
Percent Purity 97%
Chemical Name or Material 2,4-Dichloro-5-fluorobenzonitrile
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only