accessibility menu, dialog, popup

UserName

FNClCl

CAS RN 128593-93-1

IUPAC Name: 2,4-dichloro-5-fluorobenzonitrile
Synonyms: 2,4-dichloro-5-fluorobenzonitrile
Molecular Weight (g/mol): 190
Molecular Formula: C7H2Cl2FN
InChi Key: XZBZWBDTFUFZSU-UHFFFAOYSA-N
SMILES: FC1=CC(C#N)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results