missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromoanisole 98.0+%, TCI America™

Catalog No. D186325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D186325G 25 g
D1863250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D186325G Supplier TCI America Supplier No. D186325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21702-84-1
Molecular Formula C7H6Br2O
Molecular Weight (g/mol) 265.932
MDL Number MFCD00000079
InChI Key XGXUGXPKRBQINS-UHFFFAOYSA-N
PubChem CID 27011
IUPAC Name 2,4-dibromo-1-methoxybenzene
SMILES COC1=C(C=C(C=C1)Br)Br

Specifications

Melting Point 63°C
Boiling Point 101°C
Quantity 25 g
Synonym 2,4-dibromoanisole, benzene, 2,4-dibromo-1-methoxy, benzene,2,4-dibromo-1-methoxy, pubchem3847, acmc-209fnf, maybridge1_003667, chemwish ic06728, 2,4-dibromo-1-methoxy-benzene, xgxugxpkrbqins-uhfffaoysa, #
Formula Weight 265.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dibromoanisole