accessibility menu, dialog, popup

UserName

H3COBrBr

CAS RN 21702-84-1

IUPAC Name: 2,4-dibromo-1-methoxybenzene
Synonyms: 2,4-dibromo-1-methoxybenzene
Molecular Weight (g/mol): 265.93
Molecular Formula: C7H6Br2O
InChi Key: XGXUGXPKRBQINS-UHFFFAOYSA-N
SMILES: COC1=CC=C(Br)C=C1Br
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)