missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-6-nitroaniline, 99%

CAS: 827-23-6 | C6H4Br2N2O2 | 295.918 g/mol

Supplier:  Thermo Scientific Chemicals L2033314

 View more versions of this product

Catalog No. AAL2033314


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Dibromo-6-nitroaniline
129°C to 131°C
MFCD00012107
2805574
OBTUHOVIVLDLLD-UHFFFAOYSA-N
2,4-dibromo-6-nitroaniline
70009
99%
827-23-6
C6H4Br2N2O2
25 g
benzenamine, 2,4-dibromo-6-nitro, 2,4-dibromo-6-nitrophenylamine, 2,4-dibromo-6-nitrobenzenamine, acmc-209pq0, 2-nitro-4,6-dibromoaniline, 4,6-dibromo-2-nitroaniline, 2,4-dibromo-6-nitro-aniline, 2,4-dibromo-6-nitro-phenylamine, aniline, 2,4-dibromo-6-nitro, benzenamine,2,4-dibromo-6-nitro
C1=C(C=C(C(=C1Br)N)[N+](=O)[O-])Br
295.918
295.92

Chemical Identifiers

827-23-6
295.918
OBTUHOVIVLDLLD-UHFFFAOYSA-N
70009
C1=C(C=C(C(=C1Br)N)[N+](=O)[O-])Br
C6H4Br2N2O2
MFCD00012107
benzenamine, 2,4-dibromo-6-nitro, 2,4-dibromo-6-nitrophenylamine, 2,4-dibromo-6-nitrobenzenamine, acmc-209pq0, 2-nitro-4,6-dibromoaniline, 4,6-dibromo-2-nitroaniline, 2,4-dibromo-6-nitro-aniline, 2,4-dibromo-6-nitro-phenylamine, aniline, 2,4-dibromo-6-nitro, benzenamine,2,4-dibromo-6-nitro
2,4-dibromo-6-nitroaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H373-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only