missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-1-naphthol 98.0+%, TCI America™

Catalog No. D01955G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D01955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D01955G Supplier TCI America Supplier No. D01955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2050-49-9
Molecular Formula C10H6Br2O
Molecular Weight (g/mol) 301.965
MDL Number MFCD00059130
InChI Key PSGUDVJPEWTBRM-UHFFFAOYSA-N
PubChem CID 94878
IUPAC Name 2,4-dibromonaphthalen-1-ol
SMILES C1=CC=C2C(=C1)C(=CC(=C2O)Br)Br

Specifications

Melting Point 107°C
Quantity 5 g
Formula Weight 301.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dibromo-1-naphthol